Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCK2 Monoclonal Antibody (OTI4H7), TrueMAB™, OriGene
SDP

Catalog No. 501683791
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-3791 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-3791 Supplier OriGene Supplier No. TA811057
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NCK1 is one of the adaptor proteins which mediate specific protein-protein interactions in signaling processes. Adaptor proteins usually contain several domains like SH2 (Src homology 2) and SH3 which allow specific interactions with other specific proteins. NCK1 and NCK2 showing high sequence identity (68%) have three SH3 domains and a C-terminal SH2 domain. Both of them bind receptor tyrosine kinases such as PDGFR and other tyrosine phosphorylated proteins via their SH2 domains. Various molecules which interact with SH domains of Nck and regulate signaling process of actin cytoskeleton reorganization have been identified. Ncks are thought to have important functions in the development of mesodermal structures during embryogenesis, linked to a role in cell movement and cytoskeletal reorganization. Nck also have a function in modulating mRNA translation at the level of initiation by interacting eukayotic initiation factor 2 (eIF2). Under the stressed conditions, protein synthesis is reduced by inhibiting the activity of eIF2 through the phosphorylation, transiently inhibiting recycling of eIF2 into its active form.

Specifications

Antigen NCK2
Applications Western Blot
Classification Monoclonal
Clone OTI4H7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene NCK2
Gene Accession No. O43639
Gene Alias GRB4, NCKbeta
Gene Symbols NCK2
Host Species Mouse
Immunogen Full length human recombit protein of human NCK2 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8440
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.