Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NCoR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA1843
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA1843 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA1843 Supplier Invitrogen™ Supplier No. PA1843
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-843 detects silencing mediator of retinoic acid and thyroid hormone receptor (SMRT) from human and mouse cells. PA1-843 has been successfully used in Western blot, ICC/IF and immunohistochemistry (paraffin) procedures. By Western blot, this antibody detects an ~250 kDa protein representing SMRT from HeLa cell nuclear extracts. The PA1-843 antigen is His-tagged fusion protein corresponding to amino acid residues 95-565 from human SMRT.

Steroid and thyroid hormones and retinoic acid regulate a complex array of gene expression activity via intracellular receptor transcription factors belonging to the ligand-dependent nuclear receptor superfamily. Adding to the complexity of function of these transcription factors are associated proteins known as coactivators and corepressors which, as their names suggest, enhance or depress transcription activity of the nuclear receptor with which they associate. Silencing mediator of retinoic acid and thyroid hormone receptor (SMRT) and nuclear receptor corepressor (N-CoR) are related transcriptional corepressors which contain two distinct domains capable of interacting with unliganded nuclear receptors to repress their basal transcriptional activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCoR2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene NCOR2
Gene Accession No. Q9WU42, Q9Y618
Gene Alias CTG repeat protein 26; CTG26; N-CoR; NCOR2; N-CoR2; nuclear receptor corepressor; nuclear receptor corepressor 2; nuclear receptor co-repressor 2; silencing mediator for retinoid and thyroid hormone receptors; silencing mediator of retinoic acid and thyroid hormone receptor; silencing mediator of retinoid acid and thyroid hormone receptor; SMAP270; SMRT; SMRTe; SMRTE-tau; T3 receptor-associating factor; Thyroid-, retinoic-acid-receptor-associated corepressor; TNRC14; TRAC; TRAC1; TRAC-1
Gene Symbols NCOR2
Host Species Rabbit
Immunogen His-tagged fusion protein corresponding to amino acid residues 95-565 from human SMRT.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20602, 9612
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.