Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ β-Ncotinamide Adenine Dinucleotide Reduced, Disodium Salt

Catalog No. MP021011681 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021011681 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP021011681 Supplier MP Biomedicals™ Supplier No. 02101168.1

Involved in a wide range of enzyme catalyzed oxidation reduction reactions.

Applications: β-Nicotinamide adenine dinucleotide (NAD+) and β-Nicotinamide adenine dinucleotide, reduced (NADH) comprise a coenzyme redox pair (NAD+:NADH) involved in a wide range of enzyme catalyzed oxidation reduction reactions. In addition to its redox function, NAD+/NADH is a donor of ADP-ribose units in ADP-ribosylaton (ADP-ribosyltransferases; poly(ADP-ribose) polymerases ) reactions and a precursor of cyclic ADP-ribose (ADP-ribosyl cyclases). Many metabolites and enzymes of biological interest are present in tissues at low concentrations. With the use of β-NADH as a cofactor and several enzymes in a multistep system, known as enzyme cycling, much greater sensitivity for detection of these components is achieved. β-NADH is fluorescent; whereas, β-NAD+ is not. This difference in fluorescence provides a sensitive fluorescent measurement of the oxidized or reduced pyridine nucleotides at concentrations down to 10−7 M. Discussion of optimizing the fluorescence intensity and identification of interfering substances has been reported.
Product Description: β-NADH, a pyridine nucleotide and biologically active form of nicotinic acid, is a coenzyme necessary for the catalytic reaction of certain enzymes. β-NAD+ is a carrier for hydride ion, forming β-NADH. The hydride ion is enzymatically removed from a substrate molecule by the action of dehydrogenases such as, malic dehydrogenase and lactic dehydrogenase. These enzymes catalyze the reversible transfer of a hydride ion from malate or lactate to β-NAD+, forming the reduced product, β-NADH. Unlike β-NAD+, which has no absorbance at 340 nm, β-NADH absorbs at 340 nm. The increase in absorbance (with the formation of β-NADH) or the decrease in absorbance (with the formation β-NAD+) is the basis for measurement of activity of many enzymes at 340 nm.
Biochem/physiol Actions: NADH is a coenzyme that functions as a regenerating electron donor in catabolic processes including glycolysis, beta-oxidation and the citric acid cycle (Krebs cycle, TCA cycle). It participates in cell signaling events as well, for example as a substrate for the poly (ADP-ribose) polymerases (PARPs) during the DNA damage response. The NAD+/NADH dependent sirtuins play key roles in stress responses during events involving energy metabolism, with implications in cancer biology, diabetes and neurodegenerative disease. As a reagent, NADH can be used in enzyme cycling assays to amplify detection of activity of biologically relevant enzymes or metabolites present in low concentrations.
TRUSTED_SUSTAINABILITY

Specifications

Color White
pH >
For Use With (Application) Coenzyme
Content And Storage Store at -0 °C. Store Desiccated. Store Under Nitrogen. Protect from light.
Quantity 100 mg
Product Type Biochemicals
Molecular Weight (g/mol) 709.4
Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.