Learn More
Cayman Chemical -ncrostatIn-2 10mg
SDP
Supplier:  Cayman Chemical 2092410 MG
Encompass_Preferred
A RIPK1 inhibitor; 6 mg/kg/day prevents TNF-induced mortality in a murine model of SIRS; decreases irradiation-induced LDH release and cell death in murine embryonic Cyt c-/- cells; 1.6 mg/kg/day prevents angiotensin II-induced elastin degradation and aortic inflammation in a murine AAA model
Catalog No. 50-298-5378