Learn More
Selleck Chemical LLC NCT-503 5mg 1916571-90-8
SDP
Supplier:  Selleck Chemical LLC S86195MG
Encompass_Preferred
NCT-503 is a phosphoglycerate dehydrogenase (PHGDH) inhibitor with the IC50 value of 2.5 ?M. It is inactive against a panel of other dehydrogenases and shows minimal cross-reactivity in a panel of 168 GPCRs. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8165
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.