Learn More
                                    Medchemexpress LLC HY-101842 25mg , ND-646 CAS:1434639-57-2 Purity:>98%
Supplier: Medchemexpress LLC HY10184225MG
            HY-101842 25mg ND-646 CAS: 1434639-57-2 ND-646 is an orally bioavailable and steric inhibitor of acetyl-CoA carboxylase (ACC) with IC50s of 3.5 nM and 4.1 nM for recombinant hACC1 and hACC2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.