Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ndelta-Boc-L-ornithine, 98%

Catalog No. AAH6277403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6277403 1 g
AAH6277406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6277403 Supplier Thermo Scientific Chemicals Supplier No. H6277403
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 13650-49-2 | C10H20N2O4 | 232.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13650-49-2
Molecular Formula C10H20N2O4
Molecular Weight (g/mol) 232.28
MDL Number MFCD00037219
InChI Key GLZZMUULAVZVTA-ZETCQYMHSA-N
Synonym h-orn boc-oh, n-delta-boc-l-ornithine, boc-l-orn-oh, s-2-amino-5-tert-butoxycarbonyl amino pentanoic acid, ndelta-boc-l-ornithine, n-, a-boc-l-ornithine, l-ornithine, n5-1,1-dimethylethoxy carbonyl, n-epsilon-t-butyloxycarbonyl-l-ornithine, n∼5∼-tert-butoxycarbonyl-l-ornithine, s-2-amino-5-tert-butoxycarbonylamino-pentanoic acid
PubChem CID 6993436
IUPAC Name (2S)-2-amino-5-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
SMILES CC(C)(C)OC(=O)NCCCC(C(=O)O)N

Specifications

Quantity 1 g
Formula Weight 232.28
Percent Purity 98%
Chemical Name or Material Ndelta-Boc-L-ornithine
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.