Learn More
Med Vet International BD 305761 - Eclipse Safety Needle 21 x 1 1/2" 100/Box
Supplier: Med Vet International 305765
The BD Eclipse needle is a pivoting-shield needle for efficient safety during activation. The needle is bevel oriented to the safety cover to facilitate low-angle injections.Features and benefitsThe BD Eclipse needle provides streamlined and effective injection safety.Activation that requires virtually no change in clinical technique for easy integration.Designed to specifically reduce the risk of accidental needlestick injuries.Allows for shielding of the needle tip immediately after injection, with a single finger stroke.Audible click confirms activation to support your safety.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.