Learn More
Med Vet International McKesson Hypodermic Needle, 31 Gauge, 5/16 Inch Length, 16-N8MMPA, 100/Box
Supplier:  Med Vet International 1118153BX
Encompass_Preferred
Insulin Pen Needle McKesson Prevent 31 Gauge 5/16 Inch Length Safety Shield Automatic needle lock-out; appearance of red stripes confirms that the needle has been used and is ready to be disposed of safely Reduces user discomfort due to thinner needle (31G) Viewing window allows easy visibility to needle tip during priming to prevent unnecessary drug waste Wing-shaped design for comfortable and secure grip Regular bevel Stainless steel needle is a 5/16 inch in length
Catalog No. 50-304-3739
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.