Learn More
Med Vet International NDLE19GX1.5BLUNT ALUMEXEL
Supplier:  Med Vet International 26474
Encompass_Preferred
The Exel International 26474 aluminum hub blunt needles have a luer lock design and are sterile, single use, and individually packaged. They have a stainless-steel laser welded non lubricated cannula and have a 2 piece polypropylene needle cap and sheath.
Catalog No. 50-255-9740
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.