Learn More
Medi Dose Inc Eps Inc NeedleSafe II Uncapper/Recapper (For Lab R&D usage)
SDP
Supplier:  Medi Dose Inc Eps Inc NS200
Encompass
NeedleSafe II NeedleSafe II needle uncapper/recapper and syringe holder minimizes the risk of needle stick injury to all dispensing and administering personnel. It requires only a single hand for uncapping and recapping procedures. Complying with all OSHA regulations, NeedleSafe II has a Universal Gripper Core so it can even hold phlebotomy tubes and catheter caps. It's durable and reusable...reducing medication waste and its associated disposal costs. NeedleSafe II occupies very little workspace. It's easily mounted on virtually any surface...horizontal or vertical. You can place one at each patient's bedside or on every medication cart.
Catalog No. NC2703873
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.