Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ NDSB-195, Calbiochem™,

Catalog No. 48000125GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
48-000-125GM 25 g Glass Bottle
48-000-15GM 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 48-000-125GM Supplier MilliporeSigma™ Supplier No. 48000125GM
Add to Cart
Add to Cart

Chemical Identifiers

CAS 160255-06-1
Molecular Formula C7H17NO3S
Molecular Weight (g/mol) 195.277
InChI Key NNCRHRDBFDCWPA-UHFFFAOYSA-N
Synonym Dimethylethylammonium Propane Sulfonate
PubChem CID 5221296
IUPAC Name 3-[ethyl(dimethyl)azaniumyl]propane-1-sulfonate
SMILES CC[N+](C)(C)CCCS(=O)(=O)[O-]

Specifications

Color White
Quantity 25 g
Packaging Glass Bottle
Percent Purity ≥95% (TLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.