Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDST3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563262
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563262 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563262 Supplier Invitrogen™ Supplier No. PA563262
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TSEKSVQSFQ LKGFPFSIYG NLAVKDCCIN PHSPLIRVTK SSKLEKGSLP GTD Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 81%.

This gene encodes a member of the heparan sulfate/heparin GlcNAc N-deacetylase/ N-sulfotransferase family. The encoded enzyme is a type II transmembrane protein that resides in the Golgi apparatus. This monomeric bifunctional enzyme catalyzes the N-deacetylation and N-sulfation of N-acetylglucosamine residues in heparan sulfate and heparin, which are the initial chemical modifications required for the biosynthesis of the functional oligosaccharide sequences that define the specific ligand binding activities of heparan sulfate and heparin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDST3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NDST3
Gene Accession No. O95803
Gene Alias Bifunctional heparan sulfate N-deacetylase/N-sulfotransferase 3; GlcNAc N-deacetylase/ N-sulfotransferase 3; glucosaminyl N-deacetylase/N-sulfotransferase 3; Heparan sulfate N-deacetylase 3; Heparan sulfate N-sulfotransferase 3; hNDST-3; HSST3; N-deacetylase and N-sulfotransferase 3; N-deacetylase/N-sulfotransferase (heparan glucosaminyl) 3; N-deacetylase/N-sulfotransferase 3; NDST3; NDST-3; N-heparan sulfate sulfotransferase 3; N-HSST 3; UNQ2544/PRO4998
Gene Symbols NDST3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NDST3. Recombinant protein control fragment (Product #RP-95670).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9348
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.