Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDUFA13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106926
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106926 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106926 Supplier Invitrogen™ Supplier No. PA5106926
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NDUFA13.

A novel gene associated with Retinoid-IFN-induced Mortality (GRIM) GRIM-19 gene was identified. Antisense expression of GRIM-19 confers a strong resistance against IFN/RA-induced death by reducing the intracellular levels of GRIM-19 protein. Overexpression of GRIM-19 enhances cell death in response to IFN/RA. GRIM-19 is primarily a nuclear protein whose expression is induced by the IFN/RA combination. These data indicate that GRIM-19 is a novel cell death-regulatory molecule.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFA13
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Ndufa13
Gene Accession No. Q9ERS2, Q9P0J0
Gene Alias 2700054G14Rik; AU022060; B16.6; CDA016; Cell d; Cell death regulatory protein GRIM-19; cell death-regulatory protein GRIM19; CGI-39; CI-B16.6; complex I B16.6 subunit; complex I-B16.6; FLJ58045; FLJ59191; gene associated with retinoic and IFN-induced mortality 19 protein; gene associated with retinoic and interferon-induced mortality 19 protein; Gene associated with retinoic-interferon-induced mortality 19 protein; genes associated with retinoid-IFN-induced mortality 19; GRIM19; GRIM-19; LOC100911483; mitochondrial NADH:ubiquinone oxidoreductase B16.6 subunit; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 13; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 13; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 13-like; NADH:ubiquinone oxidoreductase subunit A13; NADH-ubiquinone oxidoreductase B16.6 subunit; Ndufa13; YjeF N-terminal domain-containing protein 3; YjeF_N3; Yjefn3
Gene Symbols Ndufa13, Ndufa13-ps1
Host Species Rabbit
Immunogen A synthesized peptide derived from human NDUFA13(Accession Q9P0J0), corresponding to amino acid residues M1-L43.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100911483, 314759, 51079, 67184
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.