Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDUFA3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561390
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561390 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561390 Supplier Invitrogen™ Supplier No. PA561390
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PYFKYSVMIN KATPYNYPVP VRDDGNMPDV PSHPQDPQGP SLEWLKKL Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 83%.

Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFA3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NDUFA3
Gene Accession No. O95167
Gene Alias 1010001M12Rik; 1700022J01Rik; B9; CI-B9; complex I B9 subunit; complex I-B9; NADH dehydrogenase; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 3; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 3, 9kDa; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 3; NADH:ubiquinone oxidoreductase subunit A3; NADH-ubiquinone oxidoreductase B9 subunit; ndufa3; zgc:100908
Gene Symbols NDUFA3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NDUFA3. Recombinant protein control fragment (Product #RP-99164).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4696
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.