Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDUFA4L2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120264
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120264 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120264 Supplier Invitrogen™ Supplier No. PA5120264
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: 293T, Jurkat, HepG2, HeLa, Mouse brain, Mouse kidney, Rat brain. Immunogen sequence: MAGASLGARF YRQIKRHPGI IPMIGLICLG MGSAALYLLR LALRSPDVCW DRKNNPEPWN RLSPNDQYKF LAVSTDYKKL KKDRPDF.

NDUFA4L2 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFA4L2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene NDUFA4L2
Gene Accession No. Q4FZG9, Q9NRX3
Gene Alias BC064011; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 4-like 2; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 4-like 2; NADH:ubiquinone oxidoreductase MLRQ subunit homolog; NADH-ubiquinone oxidoreductase MLRQ subunit homolog; NDUFA4, mitochondrial complex associated like 2; NDUFA4L2; NUOMS
Gene Symbols NDUFA4L2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-87 of human NDUFA4L2 (NP_064527.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100362331, 407790, 56901
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.