Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDUFAF4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589771
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589771 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589771 Supplier Invitrogen™ Supplier No. PA589771
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MGALVIRGIR NFNLENRAER EISKMKPSVA PRHPSTNSLL REQISLYPEV KGEIARKDEK LLSFLKDVYV DSKDPVSSLQ VKAAETCQEP KEFRLPKDHH FDMINIKSIP KGKISIVEAL TLLNNHKLFP ETWTAEKIMQ EYQLEQKDVN SLLKYFVTFE VEIFPPEDKK; Positive Samples: U-87MG, HeLa, K-562; Cellular Location: Lipid-anchor, Membrane, Mitochondrion.

NADH:ubiquinone oxidoreductase (complex I) catalyzes the transfer of electrons from NADH to ubiquinone (coenzyme Q) in the first step of the mitochondrial respiratory chain, resulting in the translocation of protons across the inner mitochondrial membrane. This gene encodes a complex I assembly factor. Mutations in this gene are a cause of mitochondrial complex I deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFAF4
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene NDUFAF4
Gene Accession No. Q9D1H6, Q9NQR8, Q9P032
Gene Alias 1110007M04Rik; 3000003G13Rik; AW214064; bA22L21.1; C6orf66; hormone-regulated proliferation associated protein 20; hormone-regulated proliferation-associated 20 kDa protein; hormone-regulated proliferation-associated protein of 20 kDa; Hormone-regulated proliferation-associated protein of 20 kDa homolog; hormone-regulated proliferation-associated protein, 20 kDa; Hrpap20; HSPC125; My013; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, assembly factor 4; NADH dehydrogenase (ubiquinone) complex I, assembly factor 4; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 4; NADH:ubiquinone oxidoreductase complex assembly factor 4; Ndufaf4; Protein HRPAP20
Gene Symbols NDUFAF4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-170 of human NDUFAF4 (NP_0548841).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29078, 362495, 68493
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.