Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDUFB10 Monoclonal Antibody (OTI4F1)
GREENER_CHOICE

Catalog No. PIMA526082
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526082 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526082 Supplier Invitrogen™ Supplier No. MA526082
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NDUFB10 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

NDUFB10 is an accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone. Complex I is composed of 45 different subunits.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFB10
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI4F1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NDUFB10
Gene Accession No. O96000, Q9DCS9
Gene Alias 0610011B04Rik; 22kDa; CI PDSW; CI-PDSW; Complex I PDSW; complex I PDSW subunit; complex I-PDSW; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 10; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 10, 22kDa; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 10; NADH ubiquinone oxidoreductase PDSW subunit (RH 16p13.3); NADH:ubiquinone oxidoreductase subunit B10; NADH-ubiquinone oxidoreductase PDSW subunit; NDUFB 10; NDUFB10; PDSW
Gene Symbols NDUFB10
Host Species Mouse
Immunogen Full length human recombinant protein of NDUFB10 produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4716, 479887, 681418, 68342
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.