Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDUFB9 Monoclonal Antibody (OTI4F8), TrueMAB™, OriGene
SDP

Catalog No. 501673602
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-3602 100 μL
50-254-1465 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-167-3602 Supplier OriGene Supplier No. TA502553
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LYRM3, also known as NADH dehydrogenase (ubiquinone) 1 beta subcomplex 9 (NDUFB9), is a ubiquitously expressed LYR-motif containing protein. It has been suggested to be a candidate gene for the branchio-oto-renal (BOR) syndrome, which is characterized by branchial and renal abnormalities and heredity deafness disorders. Other than its LYR-motif, LYRM3 appears to have no functional or structural relationship to either LYRM1 or LYRM2.

Specifications

Antigen NDUFB9
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4F8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene NDUFB9
Gene Accession No. Q9CQJ8, Q9Y6M9
Gene Alias B22, CI-B22, LYRM3, UQOR22
Gene Symbols Ndufb9
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 3-179 of human NDUFB9 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 299954, 4715, 475094, 66218
Target Species Canine, Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.