Learn More
Invitrogen™ NDUFS2 Recombinant Rabbit Monoclonal Antibody (JE65-29)

Description
NDUFS2 Recombinant Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | NDUFS2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | JE65-29 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | Ndufs2 |
| Gene Accession No. | O75306, Q641Y2, Q91WD5 |
| Gene Alias | AL033311; CI-49; CI-49kD; comp; complex 1, mitochondrial respiratory chain, 49-KD subunit; complex I 49kDa subunit; complex I-49kD; EMBL:AAH82067.1}; NADH dehydrogenase (ubiquinone) Fe-S protein 2; NADH dehydrogenase (ubiquinone) Fe-S protein 2, 49kDa (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] iron-sulfur protein 2, mitochondrial; NADH:ubiquinone oxidoreductase core subunit S2; NADH-coenzyme Q reductase; NADH-ubiquinone oxidoreductase 49 kDa subunit; NADH-ubiquinone oxidoreductase NDUFS2 subunit; Ndufs2; ndufs2 {ECO:0000312; unnamed protein product |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.