Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Necrostatin-1

Catalog No. aaj65341fpl
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ65341FPL 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ65341FPL Supplier Thermo Scientific Chemicals Supplier No. J65341FPL
Only null left
Add to Cart

CAS: 4311-88-0 | C13H13N3OS | 259.327 g/mol

Necrostatin-1 functions as an allosteric inhibitor of the death domain receptor-associated adaptor kinase RIP (RIP1) in the necroptosis pathway. While affecting this distinct cell death pathway, Necrostatin-1 does not present any perturbation of the Fas/TNFR triggered canonical apoptosis cascade.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Necrostatin-1 functions as an allosteric inhibitor of the death domain receptor-associated adaptor kinase RIP (RIP1) in the necroptosis pathway. While affecting this distinct cell death pathway, Necrostatin-1 does not present any pe
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4311-88-0
Molecular Formula C13H13N3OS
Molecular Weight (g/mol) 259.327
MDL Number MFCD00056916
InChI Key TXUWMXQFNYDOEZ-UHFFFAOYSA-N
Synonym necrostatin-1, mth-dl-tryptophan, nec-1, necrostatin 1, 5-1h-indol-3-ylmethyl-3-methyl-2-thioxo-4-imidazolidinone, mth-trp, methyl-thiohydantoin-tryptophan, necroptotic inhibitor, nec-1, 5-1h-indol-3-ylmethyl-3-methyl-2-sulfanylideneimidazolidin-4-one, 4-imidazolidinone, 5-1h-indol-3-ylmethyl-3-methyl-2-thioxo
PubChem CID 2828334
IUPAC Name 5-(1H-indol-3-ylmethyl)-3-methyl-2-sulfanylideneimidazolidin-4-one
SMILES CN1C(=O)C(NC1=S)CC2=CNC3=CC=CC=C32

Specifications

Color Yellow
Quantity 20 mg
Sensitivity Light sensitive
Solubility Information Soluble in DMSO (10-25mg/ml),100% ethanol (6mg/ml),and methanol (5mg/ml).
Formula Weight 259.33
Physical Form Powder
Chemical Name or Material Necrostatin-1
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.