accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4311-88-0

IUPAC Name: 5-[(1H-indol-3-yl)methyl]-3-methyl-2-sulfanylideneimidazolidin-4-one
Synonyms: 5-(1H-indol-3-ylmethyl)-3-methyl-2-sulfanylideneimidazolidin-4-one
Molecular Weight (g/mol): 259.33
Molecular Formula: C13H13N3OS
InChi Key: TXUWMXQFNYDOEZ-UHFFFAOYNA-N
SMILES: CN1C(=S)NC(CC2=CNC3=CC=CC=C23)C1=O
Form 
  • (1)
Product Type 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results