Learn More
Medchemexpress LLC HY-100573 10mg , Necrosulfonamide CAS:1360614-48-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10057310MG
Encompass_Preferred
HY-100573 10mg Necrosulfonamide CAS: 1360614-48-7 Necrosulfonamide is a necroptosis inhibitor acting by selectively targeting the mixed lineage kinase domain-like protein (MLKL). Necrosulfonamide prevents MLKL-RIP1-RIP3 necrosome complex from interacting with its downstream effectors. MLKL is a critical substrate of RIP3 during the induction of necrosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9416
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.