Learn More
Enzo Life Sciences Necrox-5 . methanesulfonate, (1mg)
SDP
Supplier:  Enzo Life Sciences ALX430167M001
Encompass_Preferred
Necrosis inhibitor. Cell permeable necrosis inhibitor with antioxidant activity. It localizes mostly in the mitochondria. Selectively blocks oxidative stress-induced necrotic cell death (0.1μM NecroX™-5 prevented ~50% cell death in H9C2 cells exposed to 400μM t-BuOOH for 2 hours LDH assay used). Does not protect against staurosporine or etoposide-induced apoptosis. Protects cells against cold shock, hypoxia and oxidative stress in vitro, as well as CCl4 -induced acute liver injury and chronic liver fibrosis in rodent models. Alternative Name: [5-(1,1-Dioxo-thiomorpholin-4-ylmethyl)-2-phenyl-1H-indol-7-yl]-(tetrahydro-pyran-4-ylmethyl)-amine. Formula: C25H31N3O3S. 2 CH4O3S. MW: 645.8. Purity: ≥95% (HPLC). Appearance: Off-white to light pink solid. Solubility: Soluble in DMSO. Long Term Storage: -20°C
Catalog No. NC0483443
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.