Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nectin 1 Monoclonal Antibody (CK8)
GREENER_CHOICE

Catalog No. 375900
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-590-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 37-590-0 Supplier Invitrogen™ Supplier No. 375900
  • $479.00
    $359.65 / Each of 1

    Save $119.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Nectin 1 Monoclonal Antibody for ELISA, Flow, IP, Western Blot

This gene encodes an adhesion protein that plays a role in the organization of adherens junctions and tight junctions in epithelial and endothelial cells. The protein is a calcium(2+)-independent cell-cell adhesion molecule that belongs to the immunoglobulin superfamily and has 3 extracellular immunoglobulin-like loops, a single transmembrane domain, and a cytoplasmic region. This protein acts as a receptor for glycoprotein D of herpes simplex viruses 1 and 2, and pseudorabies virus and mediates viral entry into epithelial and neuronal cells. Mutations in this gene cause cleft lip and palate/ectodermal dysplasia 1 syndrome as well as non-syndromic cleft lip with or without cleft palate. Alternative splicing results in multiple transcript variants encoding proteins with distinct C-termini.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nectin 1
Applications ELISA, Flow Cytometry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone CK8
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene Nectin1
Gene Accession No. Q15223, Q9JKF6
Gene Alias AI835281; AW549174; CD111; CLPED1; ectodermal dysplasia 4 (Margarita Island type); ED4; herpes simplex virus type 1 sensitivity; herpes virus entry mediator C; Herpesvirus entry mediator C; herpesvirus Ig-like receptor; HIgR; HV1S; Hvec; nectin 1; nectin cell adhesion molecule 1; nectin); Nectin1; nectin-1; nectin-1 alpha; nectin-1 delta; OFC7; poliovirus receptor-like 1; poliovirus receptor-related 1; poliovirus receptor-related 1 (herpesvirus entry mediator C; poliovirus receptor-related 1 (herpesvirus entry mediator C); poliovirus receptor-related 1 (herpesvirus entry mediator C; nectin); poliovirus receptor-related protein 1; PRR; PRR1; PVRL1; PVRR; PVRR1; SK-12
Gene Symbols Nectin1
Host Species Mouse
Immunogen Recombinant human Nectin-1 protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192183, 5818, 58235
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.