Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nectin 1 Monoclonal Antibody (R1.302)
GREENER_CHOICE

Catalog No. PIMA528594 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528594 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528594 Supplier Invitrogen™ Supplier No. MA528594
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

This antibody recognizes an extracellular epitope on CD111 (also known as Nectin 1), a 75 kDa type I transmembrane glycoprotein broadly expressed on endothelial cells, epithelial cells, neuronal cells, megakaryocytes, and CD34-positive stem cells.

This gene encodes an adhesion protein that plays a role in the organization of adherens junctions and tight junctions in epithelial and endothelial cells. The protein is a calcium(2+)-independent cell-cell adhesion molecule that belongs to the immunoglobulin superfamily and has 3 extracellular immunoglobulin-like loops, a single transmembrane domain, and a cytoplasmic region. This protein acts as a receptor for glycoprotein D of herpes simplex viruses 1 and 2, and pseudorabies virus and mediates viral entry into epithelial and neuronal cells. Mutations in this gene cause cleft lip and palate/ectodermal dysplasia 1 syndrome as well as non-syndromic cleft lip with or without cleft palate. Alternative splicing results in multiple transcript variants encoding proteins with distinct C-termini.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nectin 1
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone R1.302
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene Nectin1
Gene Accession No. Q15223
Gene Alias AI835281; AW549174; CD111; CLPED1; ectodermal dysplasia 4 (Margarita Island type); ED4; herpes simplex virus type 1 sensitivity; herpes virus entry mediator C; Herpesvirus entry mediator C; herpesvirus Ig-like receptor; HIgR; HV1S; Hvec; nectin 1; nectin cell adhesion molecule 1; nectin); Nectin1; nectin-1; nectin-1 alpha; nectin-1 delta; OFC7; poliovirus receptor-like 1; poliovirus receptor-related 1; poliovirus receptor-related 1 (herpesvirus entry mediator C; poliovirus receptor-related 1 (herpesvirus entry mediator C); poliovirus receptor-related 1 (herpesvirus entry mediator C; nectin); poliovirus receptor-related protein 1; PRR; PRR1; PVRL1; PVRR; PVRR1; SK-12
Gene Symbols Nectin1
Host Species Mouse
Immunogen Human CD111- transfected NIH/3T3 cells.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5818
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.