Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Nectin-2/CD112 Antibody Pair [HRP]
SDP

Catalog No. NB009901 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
15 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB009901 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB009901 Supplier Novus Biologicals™ Supplier No. NBP27943115PLATES
Add to Cart
Add to Cart

Polyclonal/Monoclonal Antibody Pair

specifically detects in Human samples. It is validated for ELISA, Sandwich ELISA.

Specifications

Assay Range 15.625 pg/mL
Assay Sensitivity 15.625-1000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes an antibody specific for Human Nectin-2/CD112.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias CD112, CD112 antigen, Herpes virus entry mediator B, Herpesvirus entry mediator B, herpesvirus entry protein B, HveB, HVEBpoliovirus receptor-like 2, nectin 2, Nectin-2, poliovirus receptor-related 2 (herpesvirus entry mediator B), poliovirus receptor-related protein 2, PRR2nectin-2, PVRR2poliovirus receptor related 2
Gene ID (Entrez) 5819
Gene Symbol NECTIN2
Host Species Rabbit
Kit Contents
  • Detection Antibody: 0.5 mg/mL rabbit anti-Nectin-2/CD112 polyclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Capture Antibody: 0.25 mg/mL of rabbit anti-Nectin-2/CD112 monoclonal antibody,
  • Standard: Each vial contains 50 ng of recombinant Nectin-2/CD112.
  • Quantity 15 Plates
    Regulatory Status RUO
    Storage Requirements Storage is content dependent.
    Target Nectin-2/CD112
    Technique 1. Capture Antibody: Dilute to a working concentration of 2.0 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.1 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute standard powder with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 1000 pg/mL is recommended.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.