Learn More
Description
Mouse anti Human CD112 antibody, clone R2-525 recognizes CD112, also known as Poliovirus Receptor Related 2, PRR2 or Nectin-2, a surface molecule originally reported to be homologous to CD155, the poliovirus receptor (Lopez et al. 1998). PRR2 expression is restricted to the myelo-monocytic and megakaryocytic lineages. PRR2 expression has also been detected on haematopoeitic progenitors expressing CD34.
Specifications
Specifications
| Antigen | Nectin 2 |
| Applications | Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation |
| Classification | Monoclonal |
| Clone | R2.525 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide |
| Gene | NECTIN2 |
| Gene Accession No. | Q92692 |
| Gene Alias | AI325026; AI987993; CD112; herpes virus entry mediator B; Herpesvirus entry mediator B; herpesvirus entry protein B; hveB; mHveB; MPH; Murine herpes virus entry protein B; murine herpesvirus entry protein B; nectin cell adhesion molecule 2; Nectin2; Nectin-2; Poliovirus receptor homolog; poliovirus receptor related 2; poliovirus receptor-like 2; poliovirus receptor-related 2; poliovirus receptor-related 2 (herpesvirus entry mediator B); poliovirus receptor-related protein 2; poliovirus sensitivity; PRR2; Pvr; Pvrl2; PVRR2; Pvs |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
