Learn More
Selleck Chemical LLC Nedaplatin S1826-10mg
SDP
Supplier:  Selleck Chemical LLC S182610MG
Encompass_Preferred
Nedaplatin(NSC 375101D) is a derivative of cisplatin and DNA damage agent for tumor colony forming units with IC50 of 94 M DMSO is not recommended to dissolve platinum-based drugs which can easily lead to drug inactivation
Catalog No. 50-310-9130