Learn More
TARGETMOL CHEMICALS INC Nedaplatin 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T241050MG
Encompass_Preferred
Also available in 10 mg, 25 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Nedaplatin (NSC-375101D) is a derivative of cisplatin and DNA damage agent for tumor colony forming units (IC50 94 uM). Containing a novel ring structure in which glycolate is bound to the platinum by a bidentate ligand, nedaplatin forms reactive platinum complexes that bind to nucleophilic groups in DNA, resulting in intrastrand and interstrand DNA cross-links, apoptosis and cell death. Purity 100%
Catalog No. 50-287-7757