Learn More
Chemie Tek NEDISERTIB - M3814 PEPOSERTIB
SDP
Supplier:  Chemie Tek CTM3814100MG
Encompass_Preferred
Nedisertib (M3814, Peposertib, MSC2490484A) is an orally bioavailable, highly potent and selective inhibitor of DNA activated protein kinase (DNA-PK) with IC50 of < 3 nM.
Catalog No. NC2165978