Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NEDL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113497
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113497 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113497 Supplier Invitrogen™ Supplier No. PA5113497
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NEDL2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

E3 ubiquitin-protein ligase that mediates ubiquitination of TP73. Acts to stabilize TP73 and enhance activation of transcription by TP73.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NEDL2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene HECW2
Gene Accession No. Q9P2P5
Gene Alias A730039N16Rik; D030049F17Rik; E3 ubiquitin-protein ligase HECW2; Gm971; hect; HECT, C2 and WW domain containing E3 ubiquitin protein ligase 2; HECT, C2 and WW domain containing E3 ubiquitin protein ligase 2, isoform 2; HECT, C2 and WW domain-containing protein 2; HECT-type E3 ubiquitin transferase HECW2; HECW2; KIAA1301; mKIAA1301; NEDD4-like E3 ubiquitin-protein ligase 2; NEDD4-related E3 ubiquitin ligase NEDL2; NEDL2
Gene Symbols HECW2
Host Species Rabbit
Immunogen Recombinant Human E3 ubiquitin-protein ligase HECW2 protein (495-641AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57520
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.