Learn More
Medchemexpress LLC Nedosiran (sodium) | 2247026-22-6 | 92.4% | 20,985.12 g/mol | C662H865F19N231O413P57S6.xNa | 5 MG

Supplier: Medchemexpress LLC HY132606A5MG
Nedosiran (sodium) is a GalNAc-conjugated double-stranded RNA (dsRNA) research material used in RNA interference studies targeting hepatic enzyme expression. Supplied as the sodium salt for analytical and preclinical research, it is characterized by high molecular weight and a specified HPLC purity.
- GalNAc-conjugated double-stranded RNA suitable for RNAi research.
- Sodium salt form for aqueous handling and formulation.
- Purity 92.4% (HPLC).
- Molecular weight about 20,985 Da.
- Commonly supplied as a 5 mg pack for analytical use.
- Store at -80°C for long-term storage or -20°C for short-term storage in solvent.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.