Learn More
MilliporeSigma™ Supelco™ Needle Cleaning Kit
Catalog No. 111010418 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Equipment):
GC Syringes
Catalog No. For Use With (Equipment)
11-101-0418 GC Syringes
1 options
Catalog No. 11-101-0418 Supplier MilliporeSigma™ Supelco™ Supplier No. 21578
Add to Cart
Edge
Add to Cart
Formulated specifically for GC syringes. Perfect for cleaning small orifices such as FID jets and syringe needles.
TRUSTED_SUSTAINABILITY
Specifications
Includes 70 mL Bottle (makes 700 mL) Cleaning Solution, 10 Wires in each of Five Sizes (0.076 to 0.306 mm O.D.), and Instructions
Product Type Cleaning Kit
Quantity 1 Ea.
Packaging Reusable Box
For Use With (Equipment) GC Syringes