Learn More
World Precision Instrument Round Handled Needle Holder, 14 cm
SDP
Supplier:  World Precision Instrument 14206
Encompass
These round-handled needle holders are a popular choice for researchers doing small animal surgeries. The knurled handle provides a secure grip on the surgical instrument and reduces the chance of it slipping during use. The round handle is also more comfortable, especially for long procedures. The round handles allow you to roll the instrument between your fingers as you are working, giving you more precise control over the maneuvering of the tip holding the surgical needle. The knurled handle let you apply the correct amount of pressure for accurate suturing. The handle is hollow, reducing the weight of the instrument to also reduce hand fatigue. The smooth jaw of the needle holder reduces the amount of trauma and tearing of delicate tissues and improves the visibility of the working area during a procedure. The tips are fine (0.3mm) for securely grasping tiny needles.
Catalog No. 50-822-274
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.