Learn More
Med Vet International Miltex Olsen-Hegar Needle Holder, Serrated Jaws, Extra Delicate, Tungsten, 4.75in, German, 8-14TC
Supplier: Med Vet International MIL814TC
Needle holder with scissors that allow you to cut and suture with the same instrument. Intended for 0 suture and finer. Serrated jaw for increased grip. Extra delicate finish. Tungsten carbide inlays provide increased durability and precision. 4.75in total length. Handcrafted with medical grade German steel.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.