Learn More
Med Vet International Safety Hypodermic Needles, 20G X 1in, 100/Box
Supplier:  Med Vet International 27417
Encompass_Preferred
Safety Hypodermic Needles, 20G X 1in, 100/Box Safety Needle Features One handed activation No solid surface required Simple finger stroke activation Protective arm keeps fingers behind the needle An audible "click" as well as tactile feel and a visual indicator confirm safety mechanism engagememt Needle remains locked inside activated cover Fits any brand Syringe
Catalog No. 50-209-2591
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.