Learn More
Neese Universal 35 Rainwear Bib Trousers, Without Safety Fly
Description
- Material: PVC / nylon
- Double stitched and tape sealed seams
- Flame, abrasion, tear and snag resistant
- Lightweight
- Bib front coverage
- With elastic suspenders
- Non-conductive snap closures that fastens suspenders onto garment
- Does not crack or peel in temperatures of -30° to +120°F (-34° to +49°C)
Specifications
Specifications
| Size | Large |
| Material | PVC |
| Item Description | Includes elastic suspenders |