Learn More
Neese Universal 35 Rainwear Bib Trousers, Without Safety Fly
SDP
Catalog No. 18816041
Change view
Click to view available options
Color:
Green
Yellow
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6X-Large
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
18-816-041 Green 3X-Large
19-020172 Yellow Small
19-020157 Yellow 2X-Large
18-816-039 Green X-Large
18-816-038 - Large
18-816-040 Green 2X-Large
18-816-042 Green 4X-Large
18-816-044 Green 6X-Large
18-816-037 - Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 18-816-041 Supplier Neese Supplier No. 35BT SP01 3X
This item is not returnable. View return policy
Provide tough protection
  • Material: PVC / nylon
  • Double stitched and tape sealed seams
  • Flame, abrasion, tear and snag resistant
  • Lightweight
  • Bib front coverage
  • With elastic suspenders
  • Non-conductive snap closures that fastens suspenders onto garment
  • Does not crack or peel in temperatures of -30° to +120°F (-34° to +49°C)
Specifications
Size 3X-Large
Material PVC
Color Green
Item Description Includes elastic suspenders