Learn More
Neese Universal 35 Rainwear Coats
SDP
Catalog No. 19412766
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Hood Type:
Attached
Detachable
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Hood Type
19-412-766 Medium Attached
19-020155 Large Attached
19-412-765 Small Attached
19-412-768 2X-Large Attached
19-412-767 X-Large Attached
19-412-769 3X-Large Attached
19-412-770 3X-Large Detachable
19-815-992 X-Large Detachable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 19-412-766 Supplier Neese Supplier No. 35001301YELM
Add to Cart
Add to Cart
This item is not returnable. View return policy
  • PVC / Nylon / PVC
  • Flame Resistant (ASTM D6413)
  • Coat style that is minimum of 48 in. long with adjustments for various sizes
  • Snap front
  • Storm flap
  • Attached hood with drawstring closure or stand up collar with snaps for hood
  • Raglan shoulder design to allow for easy movement
Specifications
Size Medium
Material Nylon, PVC
Color Yellow
Closure Type Snap, Storm Flap
Hood Type Attached
Sleeve Style Raglan