Learn More
Neese Universal 35 Rainwear Coats
Description
- PVC / Nylon / PVC
- Flame Resistant (ASTM D6413)
- Coat style that is minimum of 48 in. long with adjustments for various sizes
- Snap front
- Storm flap
- Attached hood with drawstring closure or stand up collar with snaps for hood
- Raglan shoulder design to allow for easy movement
Specifications
Specifications
| Size | 2X-Large |
| Material | Nylon, PVC |
| Color | Yellow |
| Closure Type | Snap, Storm Flap |
| Hood Type | Attached |
| Sleeve Style | Raglan |