Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Negative Control for Mouse IgG1 Ab-1, Epredia™

Catalog No. N748P Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Concentration:
1mg/mL
200μg/mL
Prediluted
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
N748P 1 mL 200μg/mL
N748PABX 200 μL 1mg/mL
N748P1 500 μL 200μg/mL
N748R7 7 mL Prediluted
N748P0 100 μL 200μg/mL
N748P1ABX 100 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. N748P Supplier Epredia Supplier No. NC748P

Provides accurate, reproducible results with the Epredia™ Negative Control for Mouse IgG1 Ab-1, Mouse Monoclonal Antibody.

This mouse monoclonal IgG1 was produced against a synthetic hapten, which is normally not present in humans or animals. Therefore, this mouse IgG1 does not react with any antigen of human or animal cells. This monoclonal IgG1 was purified from mouse ascites produced under the same experimental conditions as the other hybridoma cell lines of NeoMarkers. In order to serve as a true negative control, this IgG1 should be used under the identical conditions and at the same concentration as NeoMarkers' other purified mouse monoclonal IgG1 antibodies.

Host Species: Mouse

Clone: NCG01

Isotype: IgG1/κ

Species Reactivity: All species.

Recommended for:

Negative control for:

  • Immunohistochemistry (Frozen and formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation
  • Flow Cytometry
  • ELISA
  • Gel Supershift

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Negative Control for Mouse IgG1 Ab-1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone NCG01
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Quantity 1 mL
Regulatory Status RUO
Research Discipline Negative Controls
Primary or Secondary Primary
Target Species All species
Form Concentrated
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.