Learn More
Description
Host Species: Mouse
Clone: NCG01
Isotype: IgG1/κ
Species Reactivity: All species.
Recommended for:
Negative control for:
- Immunohistochemistry (Frozen and formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation
- Flow Cytometry
- ELISA
- Gel Supershift
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Negative Control for Mouse IgG1 Ab-1 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | NCG01 |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Quantity | 200 μL |
| Regulatory Status | RUO |
| Research Discipline | Negative Controls |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.