Learn More
TARGETMOL CHEMICALS INC BETA-ELEMONIC ACID 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T2S0618
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (beta)-Elemonic Acid (3-Oxotirucallenoic Acid) exhibits anti-inflammatory effects. Beta-Elemonic acid inhibits proliferation by inducing hypoploid cells and cell apoptosis the anticancer effects of beta-Elemonic acid are related to the MAPK signaling pathway ROS activation and glutathione depletion in human A549 lung cancer cells. Beta-Elemonic acid exhibits prolyl endopeptidase inhibitory activities. purity: 99%
Catalog No. 50-258-0439
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.