Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NEGR1 Monoclonal Antibody (OTI3G8)
GREENER_CHOICE

Catalog No. PIMA527279
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIMA527279 100 μL
1 options

Catalog No. PIMA527279

Supplier: Invitrogen™ MA527279

Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NEGR1 Monoclonal Antibody for Western Blot, IHC (P)

May be involved in cell-adhesion. May function as a trans-neural growth-promoting factor in regenerative axon sprouting in the mammalian brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NEGR1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3G8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NEGR1
Gene Accession No. Q7Z3B1
Gene Alias 5330422G01Rik; a kindred of IgLON; DMML2433; IgLON family member 4; IGLON4; Kiaa3001; KILON; kilon protein; kindred of IgLON; NEGR1; Neuronal growth regulator 1; neurotractin; Ntra; UNQ2433; UNQ2433/PRO4993
Gene Symbols NEGR1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 80-205 of NEGR1 produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 257194
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.