Learn More
Invitrogen™ Neisseria gonorrhoeae Monoclonal Antibody (GC12-351.4), Invitrogen™
Shop All Thermo Scientific Products

Description
MA1-10717 detects Neisseria gonorrhoeae. MA1-10717 has been successfully used in ELISA applications. The MA1-10717 immunogen is UV-inactivated Neisseria gonorrhoeae cells W-I, W-II, W-III.
Specifications
Specifications
| Antigen | Neisseria gonorrhoeae |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | GC12-351.4 |
| Concentration | 1.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene Alias | Gonococci; N. gonorrhoeae |
| Host Species | Mouse |
| Immunogen | UV-inactivated Neisseria gonorrhoeae cells W-I, W-II, W-III. |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.