Learn More
Creative Biomart Recombinant Japanese Encephalitis Virus ENV Antigen, His-tagged
Supplier:  Creative Biomart VAC0336
Encompass_Preferred
The E. coli derived recombinant protein contains full length Japanese Encephalitis virus ENV antigen having an Mw of 50 kDa. The protein is fused to a 6 histidines tag.
Catalog No. 50-208-5176
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.