Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Neisseria meningitidis Orf1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01676759
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-676-759 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-676-759 Supplier Invitrogen™ Supplier No. PA119444
  • $510.00
    $424.40 / Each of 1

    Save $85.60

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody reacts with Orf1 (FrpD), a 28 - 32 kDa Neisseria meningitidis outer membrane lipoprotein. Orf1 is highly conserved in a set of meningococcal strains representative of all serogroups and does not exhibit any similarity to known sequences of other organisms. Western Blot: Reducing conditions.

The genus Neisseria contains a number of species which are normal flora and pathogens of humans and animals. Of these species, the species of human origin, and particularly the pathogenic species, N. gonorrhoeae and N. meningitidis, have been studied extensively in an effort to control the infections they cause. Gonorrhea, caused by N. gonorrhoeae, is one of the most frequently reported infectious diseases in the United States and worldwide. Of the Neisseria and related species, only N. gonorrhoeae is considered always to be pathogenic i.e., cause disease. N. gonorrhoeae is not considered to be normal flora under any circumstances.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neisseria meningitidis Orf1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene Alias Meningococcus
Host Species Rabbit
Immunogen Slices containing purified FrpD250-6xHis-tagged protein were excised from 10% SDS-PAGE gels, grinded, mixed with 1 mg/ml of aluminum hydroxide in PBS.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.