Learn More
ATCC Vibrio campbellii
Supplier:  ATCC 700104
Encompass
Vibrio campbellii strain BB151 is a mutant derived from Vibrio campbellii strain BB120 (ATCC BAA-1116). This whole-genome sequenced bacterial strain produces beta-hydroxybutryl homoserine lactone (A1-1) and an unidentified autoinducer (A1-2). This strain has genotype luxA::Tn5lac.
Catalog No. 50-237-9616
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.