Learn More
ATCC Neisseria perflava
Supplier:  ATCC 14799
Encompass
Neisseria perflava strain 28 is a whole-genome sequenced bacterium with applications as a quality control strain for BBL products. This strain is propagated in a 5% CO2 atmosphere in GC medium.
Catalog No. 50-238-4620
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.